Learn More
Selleck Chemical LLC Resveratrol 500mg 501-36-0
SDP
Catalog No. 508806
Encompass_Preferred
Resveratrol has a wide spectrum of targets including cyclooxygenases(i.e. COX, IC50=1.1 ?M), lipooxygenases(LOX, IC50=2.7 ?M), kinases, sirtuins and other proteins. It has anti-cancer, anti-inflammatory, blood-sugar-lowering and other beneficial cardiovascular effects. Resveratrol induces mitophagy/autophagy and autophagy-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-880-6 Supplier Selleck Chemical LLC Supplier No. S1396500MG
Resveratrol has a wide spectrum of targets including cyclooxygenases(i.e. COX, IC50=1.1 ?M), lipooxygenases(LOX, IC50=2.7 ?M), kinases, sirtuins and other proteins. It has anti-cancer, anti-inflammatory, blood-sugar-lowering and other beneficial cardiovascular effects. Resveratrol induces mitophagy/autophagy and autophagy-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.