Learn More
BIOGEMS INTERNATIONAL INC Resveratrol 100mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 5013606100MG
Encompass
Resveratrol is a phenolic phytoalexin that has antioxidant properties and selective inhibits COX-1. It is reported to activate SIrtuin 1 and PGC-1alpha and increase the activity of SOD2. Resveratrol is an anti-inflmammatory and antiproliferative agent that interferes with carcinogenesis.
Catalog No. 50-153-0169