Learn More
Cayman Chemical RET EXrAClular DomaInhum 10ug
SDP
Catalog No. 502946335
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged RET expressed in insect cells • Amino acids: 658-1114 • MW: 77.5 kDa
Catalog No. 50-294-6335 Supplier Cayman Chemical Supplier No. 3398710 UG
Source: Active recombinant human N-terminal GST-tagged RET expressed in insect cells • Amino acids: 658-1114 • MW: 77.5 kDa