Learn More
Medchemexpress LLC HY-133553 5mg Medchemexpress, RET-IN-3 CAS:2414374-53-9 Purity:>98%
SDP
Catalog No. 502081345
Encompass_Preferred
Medchemexpress, HY-133553 5mg RET-IN-3 CAS:2414374-53-9 RET-IN-3 (compound 34) is a selective RETV804M kinase inhibitor, with an IC50 of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1345 Supplier Medchemexpress LLC Supplier No. HY1335535MG
Medchemexpress, HY-133553 5mg RET-IN-3 CAS:2414374-53-9 RET-IN-3 (compound 34) is a selective RETV804M kinase inhibitor, with an IC50 of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.