Learn More
Carnabio Usa Inc RET(M918T)/5UG/CYTOPLASMICRET
Supplier:  Carnabio Usa Inc 085085
Encompass
RET [M918T], Cytoplasmic domain, Mutant [M918T], Amino Acid 658-1114, NP_066124.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0606