Learn More
Sino Biological Recombinant Human RET Protein (R749T, GST Tag), Active
Catalog No. 502573439
Encompass_Preferred
Recombinant human RET (R749T) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3439 Supplier Sino Biological Supplier No. R0212EG50
Recombinant human RET (R749T) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.