Learn More
Carnabio Usa Inc RET(S891A)/10UG/CYTOPLASMICRE
Supplier:  Carnabio Usa Inc 0852310
Encompass
RET [S891A], Cytoplasmic domain, Mutant [S891A], Amino Acid 658-1114, NP_066124.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0969