Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RET [V804E], GST Tag Recombinant Protein

Catalog No. A33507
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33507 10 μg
A33561 100 μg
A33730 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A33507 Supplier Invitrogen™ Supplier No. A33507
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This RET V804E recombinant human protein (858-1114) was expressed in insect cells. RET V804E recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant The gene accession number for RET [V804E] is NP_066124.1.

RET receptor tyrosine kinase or ret proto-oncogene is a member of the cadherin superfamily which are cell-surface molecules that transduce signals for cell growth and differentiation. RET can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. RET signaling pathway, by regulating the development of both the nervous and lymphoid system in the gut, plays a key role in the molecular mechanisms that orchestrate intestine organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07949
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM HEPES with 5mM DTT, 15mM glutathione, 20% glycerol, 100mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 5979
Molecular Weight (g/mol) 80.5 kDa
Name Human RET [V804E], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias RET
Common Name RET (V804E Mutant)
Gene Symbol RET
Kinase Family RET Kinase Family
Protein Length 858-1114
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 858-1114
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Cytoplasmic, Mutant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.