Learn More
Eurofins Discoverx Ret (V804L), active, 10 µg. (14-758)
Supplier:  Eurofins Discoverx 14758
Encompass
N-terminal GST-tagged recombinant, human Ret amino acids 658-end containing the V804L mutation. The V804L mutation has been identified in patients with familial medullary thyroid carcinoma (FMTC) (Lombardo F. et al., (2002), JCEM, 87, 1674-1680; Lesueur F. et al, (2005), JCEM, 90, 3454-3457). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9297