Learn More
Eurofins Discoverx Ret (V804M), active, 10 µg. (14-760)
Supplier:  Eurofins Discoverx 14760
Encompass
N-terminal GST-tagged recombinant, human Ret amino acids 658 – end containing the V804M mutation. The V804M mutation has been identified in patients with familial medullary thyroid carcinoma (FMTC) (Fink et al, (1996), Int. J. Cancer, 69, 312-316; Lesueur F. et al,(2005), JCEM, 90, 3454-3457). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9301