Learn More
Carnabio Usa Inc RET(Y791F)/5UG/CYTOPLASMICRET
Supplier:  Carnabio Usa Inc 085215
Encompass
RET [Y791F], Cytoplasmic domain, Mutant [Y791F], Amino Acid 658-1114, NP_066124.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0608