Learn More
Accuflow Systems Saint-Gobain Retainer, PVDF, 1/8 in, Smooth Inner Bore
Catalog No. NC2868125
Encompass_Preferred
Saint-Gobain Pure-Fit SIB retainer provides a smooth inner-bore transition between tubing and fittings to minimize dead volume and potential leak or contamination points. Made from PVDF for chemical resistance and sterilization compatibility, the retainer is intended for bioprocess and fluid-handling applications requiring biocompatibility.
  • Provides seamless transition between tubing and fittings to minimize dead space.
  • Smooth inner bore reduces turbulence and sites for contamination.
  • PVDF construction is autoclavable and gamma stable.
  • Meets USP Class VI and relevant ISO biocompatibility criteria.
  • Nominal inner diameter 1/8 in (3.2 mm).
Catalog No. NC2868125 Supplier Accuflow Systems Supplier No. PF135125PVDFAL
Saint-Gobain Pure-Fit SIB retainer provides a smooth inner-bore transition between tubing and fittings to minimize dead volume and potential leak or contamination points. Made from PVDF for chemical resistance and sterilization compatibility, the retainer is intended for bioprocess and fluid-handling applications requiring biocompatibility.
  • Provides seamless transition between tubing and fittings to minimize dead space.
  • Smooth inner bore reduces turbulence and sites for contamination.
  • PVDF construction is autoclavable and gamma stable.
  • Meets USP Class VI and relevant ISO biocompatibility criteria.
  • Nominal inner diameter 1/8 in (3.2 mm).