Learn More
Perkin Elmer US LLC Packing Retaining Gauze - 100 Mesh, Pkg. 100
Catalog No. 509051880
Encompass_Preferred
Included in the Thermal Desorber Starter Kit (M0413541). Mesh Size: 100 x 100 Sheet Size: 12 inch x 12 inch Wire Diameter: 0.0045 inch
Catalog No. 50-905-1880 Supplier Perkin Elmer US LLC Supplier No. L4071034
Included in the Thermal Desorber Starter Kit (M0413541). Mesh Size: 100 x 100 Sheet Size: 12 inch x 12 inch Wire Diameter: 0.0045 inch