Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Retainer for 24 350mL bottle

Catalog No. 5000257014 Shop All Hach Company Products
Encompass

Retainer for (24) 350 mL bottles for use with standard portable base

Catalog No. 50-002-57014 Supplier Hach Company Supplier No. 2189
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retainer for (24) 350 mL bottles for use with standard portable base

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.