Learn More
Accuflow Systems Saint-Gobain Retainer, PVDF, 5/16 in, Pure-Fit SIB
Catalog No. NC2868128
Encompass_Preferred
Retainer for smooth inner bore (SIB) hose-barb connectors, sized for 5/16 in (8.0 mm) tubing. It secures tubing with radial compression to provide leak-resistant connections in bioprocess fluid paths. Constructed from PVDF, the retainer offers autoclave and gamma stability, low water absorption, and a high recommended working temperature suitable for USP Class VI compatible systems.
  • PVDF construction, autoclaveable and gamma stable.
  • Designed for 5/16 in (8.0 mm) tubing.
  • Provides 360° compression for leak-resistant connections.
  • Compatible with common bioprocess flexible tubing.
  • Meets USP Class VI and applicable ISO biocompatibility criteria.
  • Low water absorption (0.03%) and max recommended working temperature 275 °F (135 °C).
Catalog No. NC2868128 Supplier Accuflow Systems Supplier No. PF135312PVDFAL
Retainer for smooth inner bore (SIB) hose-barb connectors, sized for 5/16 in (8.0 mm) tubing. It secures tubing with radial compression to provide leak-resistant connections in bioprocess fluid paths. Constructed from PVDF, the retainer offers autoclave and gamma stability, low water absorption, and a high recommended working temperature suitable for USP Class VI compatible systems.
  • PVDF construction, autoclaveable and gamma stable.
  • Designed for 5/16 in (8.0 mm) tubing.
  • Provides 360° compression for leak-resistant connections.
  • Compatible with common bioprocess flexible tubing.
  • Meets USP Class VI and applicable ISO biocompatibility criteria.
  • Low water absorption (0.03%) and max recommended working temperature 275 °F (135 °C).