Learn More
Med Vet International JorNet Tubular Elastic Retainer Net - 11, J1533E, Each
Catalog No. 5000495057
Encompass_Preferred
Many indications for bandaging are a challenge because of anatomical contours and large surfaces to cover. The JorNet is a tight, but elastic mesh that fits over body shapes of any type. It remains in position to hold primary and secondary wound dressing in place. Eliminates the need for sticky tape in many cases. Features: • Reduces the need to shave fur • Can replace both adhesive tape and gauze wrap • Considerable cost savings • Sizes to fit all sizes of animals • Simply roll out and cut desired length • 25 yds. stretched in a dispensing box J1533e Net Size: #11 Medium to Large horse neck (over ears)
Catalog No. 50-004-95057 Supplier Med Vet International Supplier No. JOR1533E
Many indications for bandaging are a challenge because of anatomical contours and large surfaces to cover. The JorNet is a tight, but elastic mesh that fits over body shapes of any type. It remains in position to hold primary and secondary wound dressing in place. Eliminates the need for sticky tape in many cases. Features: • Reduces the need to shave fur • Can replace both adhesive tape and gauze wrap • Considerable cost savings • Sizes to fit all sizes of animals • Simply roll out and cut desired length • 25 yds. stretched in a dispensing box J1533e Net Size: #11 Medium to Large horse neck (over ears)