Learn More
Glass Expansion Retaining Ring for D-Torch Outer Tube
SDP
Supplier:  Glass Expansion 318082818
Encompass_Preferred
Retaining Ring for D-Torch Outer Tube. Compatible with Glass Expansion’s D-Torch for Agilent (Varian) 700-ES and Vista Axial and Radial ICP-OES (30-808-2919 and 501907026, respectively), and Nu Plasma D-Torch, 501907031.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7155