Learn More
Glass Expansion Retaining Ring for D-Torch Outer Tube
SDP
Catalog No. 502703125
Encompass_Preferred
Retaining Ring for D-Torch Outer Tube. Compatible with Glass Expansion D-Torches for Agilent 5000 SVDV/VDV/RV Series ICP-OES. Used to secure the quartz or ceramic outer tubes to the D-Torch base and inner tube.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3125 Supplier Glass Expansion Supplier No. 318083566
Retaining Ring for D-Torch Outer Tube. Compatible with Glass Expansion D-Torches for Agilent 5000 SVDV/VDV/RV Series ICP-OES. Used to secure the quartz or ceramic outer tubes to the D-Torch base and inner tube.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001