Learn More
Glass Expansion Retaining Ring for D-Torch
SDP
Catalog No. NC2328922
Encompass_Preferred
Retaining Ring for D-Torch. ONLY compatible with Glass Expansion D-Torches for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2328922 Supplier Glass Expansion Supplier No. 318082707
Retaining Ring for D-Torch. ONLY compatible with Glass Expansion D-Torches for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001