Learn More
Starna Cells Inc RETANGULAR CELL 21Q2
SDP
Supplier:  Starna Cells Inc 21Q2
Encompass_Preferred
Cell, Rectangular; Standard; Starna; w/stopper; Window and Wall Thickness: 1.25mm; Exteroir: 4.5L x 12.5W x 55mmH; Interior: 10W x 2mm long; Polished Windows: 2; Path Length: 2mm; Window Material: Spectrosil Quartz
Catalog No. NC9868622