Learn More
MSC Starrett Rectangle Steel Gage Block, 0.14 in., Grade A1, 1 ea.
Catalog No. 5000512865
Encompass
This rectangle gage block is designed for precision measurement and calibration applications, providing a reliable standard for checking and setting instruments. Constructed from durable steel, it is engineered for stability and accuracy in industrial measurement environments.
  • Manufactured from steel for long-term durability
  • Grade A1 accuracy
  • Rectangle geometry
  • Made in the USA
Catalog No. 50-005-12865 Supplier MSC Supplier No. 05555727
This rectangle gage block is designed for precision measurement and calibration applications, providing a reliable standard for checking and setting instruments. Constructed from durable steel, it is engineered for stability and accuracy in industrial measurement environments.
  • Manufactured from steel for long-term durability
  • Grade A1 accuracy
  • Rectangle geometry
  • Made in the USA