Learn More
Apexbio Technology LLC Retapamulin,10mg CAS# 224452-66-8
SDP
Catalog No. 502240453
Encompass_Preferred
Retapamulin is a pleuromutilin antibiotic [1].Retapamulin is developed for topical treatment of skin infections which is mainly caused by gram-positive organisms, β-hemolytic streptococci and coagulase-negative staphylococci. Retapamulin is especially used against those drug resistance strains. In the in vitro studies, retapamulin demonstrates potent activity and a broad spectrum. It shows excellent antimicrobial activities against S. aureus, S. epidermidis, S. saprophyticus, S. pyogenes, S. agalactiae and Viridans group streptococci with MIC50 values of 0.06μg/ml, 0.06μg/ml, 0.12μg/ml, 0.008μg/ml, 0.016μg/ml and 0.03μg/ml, respectively. Retapamulin plays its antibiotic role through binding to a unique site of the bacterial ribosome [1, 2]. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0453 Supplier Apexbio Technology LLC Supplier No. B201910
Retapamulin is a pleuromutilin antibiotic [1].Retapamulin is developed for topical treatment of skin infections which is mainly caused by gram-positive organisms, β-hemolytic streptococci and coagulase-negative staphylococci. Retapamulin is especially used against those drug resistance strains. In the in vitro studies, retapamulin demonstrates potent activity and a broad spectrum. It shows excellent antimicrobial activities against S. aureus, S. epidermidis, S. saprophyticus, S. pyogenes, S. agalactiae and Viridans group streptococci with MIC50 values of 0.06μg/ml, 0.06μg/ml, 0.12μg/ml, 0.008μg/ml, 0.016μg/ml and 0.03μg/ml, respectively. Retapamulin plays its antibiotic role through binding to a unique site of the bacterial ribosome [1, 2]. Other sizes are also available. Please inqury us for quote.