Learn More
Medchemexpress LLC HY-10210 1mg Medchemexpress, Retaspimycin (Hydrochloride) CAS:857402-63-2 Purity:>98%
SDP
Catalog No. 501967982
Encompass_Preferred
Medchemexpress, HY-10210 1mg Retaspimycin (HCl) CAS:857402-63-2 Retaspimycin Hydrochloride is a potent and water-soluble inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7982 Supplier Medchemexpress LLC Supplier No. HY102101MG
Medchemexpress, HY-10210 1mg Retaspimycin (HCl) CAS:857402-63-2 Retaspimycin Hydrochloride is a potent and water-soluble inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.