Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retaspimycin hydrochloride | 857402-63-2 | 624.17 | C31H46ClN3O8 | 1 ML
SDP

Catalog No. 5000246939
Encompass_Preferred

Retaspimycin hydrochloride is a small-molecule inhibitor of heat shock protein 90 (Hsp90) provided as a concentrated DMSO stock for research use. It exhibits submicromolar activity against HSP90 and GRP94 and is supplied with guidance on solubility and storage for experimental applications.

  • Potent Hsp90 and GRP94 inhibitor with reported EC50 of 119 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) for convenient stock preparation.
  • Molecular weight 624.17 and formula C31H46ClN3O8 for accurate dosing calculations.
  • High solubility in DMSO and documented in vivo formulation options.
  • Recommended storage at -20°C (solid) and -80°C in solution for long-term stability.

Catalog No. 50-002-46939 Supplier Medchemexpress LLC Supplier No. HY1021010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retaspimycin hydrochloride is a small-molecule inhibitor of heat shock protein 90 (Hsp90) provided as a concentrated DMSO stock for research use. It exhibits submicromolar activity against HSP90 and GRP94 and is supplied with guidance on solubility and storage for experimental applications.

  • Potent Hsp90 and GRP94 inhibitor with reported EC50 of 119 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) for convenient stock preparation.
  • Molecular weight 624.17 and formula C31H46ClN3O8 for accurate dosing calculations.
  • High solubility in DMSO and documented in vivo formulation options.
  • Recommended storage at -20°C (solid) and -80°C in solution for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.