Learn More
Medchemexpress LLC Retaspimycin hydrochloride | 857402-63-2 | 96.2% | 624.17 g/mol | C31H46ClN3O8 | 100 MG
SDP
Catalog No. 5000245825
Encompass_Preferred
Retaspimycin hydrochloride is a small-molecule inhibitor targeting heat shock protein 90 (Hsp90). It is the hydrochloride salt form and has been reported to inhibit Hsp90 and Grp94 with an EC50 of approximately 119 nM; it has been used in studies of unfolded protein response and downstream signaling pathways.
  • Potent Hsp90 and Grp94 inhibitor (EC50 ≈ 119 nM).
  • Hydrochloride salt form to improve solubility for assays.
  • Molecular formula C31H46ClN3O8; molecular weight 624.17 g/mol.
  • Available in multiple sizes and solution formats for flexible use.
  • Intended for research use only (nonclinical applications).
Catalog No. 50-002-45825 Supplier Medchemexpress LLC Supplier No. HY10210100MG
Retaspimycin hydrochloride is a small-molecule inhibitor targeting heat shock protein 90 (Hsp90). It is the hydrochloride salt form and has been reported to inhibit Hsp90 and Grp94 with an EC50 of approximately 119 nM; it has been used in studies of unfolded protein response and downstream signaling pathways.
  • Potent Hsp90 and Grp94 inhibitor (EC50 ≈ 119 nM).
  • Hydrochloride salt form to improve solubility for assays.
  • Molecular formula C31H46ClN3O8; molecular weight 624.17 g/mol.
  • Available in multiple sizes and solution formats for flexible use.
  • Intended for research use only (nonclinical applications).