Learn More
Cayman Chemical RetatrutIdesodIum salt 10mg
SDP
Catalog No. 502941996
Encompass_Preferred
A GCGR, GLP-1R, and GIP receptor agonist; induces cAMP accumulation in HEK293 cells expressing the human GCGR, GLP-1R, or GIP receptor (EC50s = 5.79, 0.775, and 0.0643 nM, respectively); decreases plasma glucose levels in an intraperitoneal glucose tolerance test in mice from 0.1-30 nmol/kg; decreases body weight, calorie intake, fat mass, and lean mass in a mouse model of diet-induced obesity; acute administration delays gastric emptying and chronic administration for 10 days decreases food intake and body weight in mice
Catalog No. 50-294-1996 Supplier Cayman Chemical Supplier No. 3974710 MG
A GCGR, GLP-1R, and GIP receptor agonist; induces cAMP accumulation in HEK293 cells expressing the human GCGR, GLP-1R, or GIP receptor (EC50s = 5.79, 0.775, and 0.0643 nM, respectively); decreases plasma glucose levels in an intraperitoneal glucose tolerance test in mice from 0.1-30 nmol/kg; decreases body weight, calorie intake, fat mass, and lean mass in a mouse model of diet-induced obesity; acute administration delays gastric emptying and chronic administration for 10 days decreases food intake and body weight in mice