Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retene, 98%, MP Biomedicals

Catalog No. ICN217893 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217893 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217893 Supplier MP Biomedicals, Inc Supplier No. 0521789301

Chemical Identifiers

CAS 483-65-8
Molecular Formula C18H18
Molecular Weight (g/mol) 234.34
MDL Number MFCD00016358
InChI Key NXLOLUFNDSBYTP-UHFFFAOYSA-N
Synonym retene, reten, 1-methyl-7-isopropylphenanthrene, 7-isopropyl-1-methylphenanthrene, phenanthrene, 1-methyl-7-1-methylethyl, 1-methyl-7-1-methylethyl phenanthrene, unii-0w2d2e1p9q, phenanthrene, 7-isopropyl-1-methyl, ccris 3180, 7-isopropyl-1-methylphenanthren
PubChem CID 10222
IUPAC Name 1-methyl-7-(propan-2-yl)phenanthrene
SMILES CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1

Specifications

Quantity 1 g
Formula Weight 234.32
Chemical Name or Material Retene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.