Learn More
Medchemexpress LLC HY-P1347A 5mg Medchemexpress, RETF-4NA (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1347A5MG
Encompass_Preferred
Medchemexpress, HY-P1347A 5mg RETF-4NA (TFA) CAS: RETF-4NA TFA, a chymase-specific substrate, is a sensitive and selective substrate for chymase when free or bound to α2M. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3053