Learn More
Electron Microscopy Sciences Double Cross Lines Reticle RET 60, 19 mm, 0.025 mm Gap
SDP
Catalog No. 501901542
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.025 mm
Catalog No. 50-190-1542 Supplier Electron Microscopy Sciences Supplier No. 6026019
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.025 mm