Learn More
Electron Microscopy Sciences Double Cross Lines Reticle RET 61, 21 mm, 0.075 mm Gap
SDP
Catalog No. 501901545
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Gap: 0.075 mm
Catalog No. 50-190-1545 Supplier Electron Microscopy Sciences Supplier No. 6026121
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Gap: 0.075 mm