Learn More
Electron Microscopy Sciences Double Cross Lines Reticle RET 62, 19 mm, 0.125 mm Gap
SDP
Catalog No. 501901546
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.125 mm
Catalog No. 50-190-1546 Supplier Electron Microscopy Sciences Supplier No. 6026219
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.125 mm