Learn More
Electron Microscopy Sciences Double Cross Lines Reticle RET 63, 19 mm, 0.50 mm Gap
SDP
Catalog No. 501901547
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.50 mm
Catalog No. 50-190-1547 Supplier Electron Microscopy Sciences Supplier No. 6026319
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 0.50 mm