Learn More
Electron Microscopy Sciences Double Cross Lines Reticle RET 64, 19 mm, 1.0 mm Gap
SDP
Catalog No. 501901549
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 1.0 mm
Catalog No. 50-190-1549 Supplier Electron Microscopy Sciences Supplier No. 6026419
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Gap: 1.0 mm