Learn More
Electron Microscopy Sciences Measuring Reticle B with Frame
SDP
Catalog No. 5034561
Encompass_Preferred
Measuring reticle for Macroscope 25, frequently used in textiles or electronics and where measurement in mils is required, 4⁄16" long, 1⁄64" divisions.
Catalog No. 50-345-61 Supplier Electron Microscopy Sciences Supplier No. 67961
Measuring reticle for Macroscope 25, frequently used in textiles or electronics and where measurement in mils is required, 4⁄16" long, 1⁄64" divisions.