Learn More
Electron Microscopy Sciences Measuring Reticle C with Frame
SDP
Catalog No. 5034560
Encompass_Preferred
Measuring reticle for Macroscope 25, calibrated in um and helpful for pesticide droplet size measurements or anywhere measurements in microns are required, 5000 microns long, with 50 micron divisions.
Catalog No. 50-345-60 Supplier Electron Microscopy Sciences Supplier No. 67962
Measuring reticle for Macroscope 25, calibrated in um and helpful for pesticide droplet size measurements or anywhere measurements in microns are required, 5000 microns long, with 50 micron divisions.