Learn More
Electron Microscopy Sciences Reticle Stage Micrometer 1 mm Scale With 100 Div
SDP
Supplier:  Electron Microscopy Sciences A9STM
Encompass_Preferred
Reticles for use with our compound and inverted light microscopes. Reticle and stage micrometer with 1 mm scale with 100 divisions for use with compound microscopes.
Catalog No. 50-342-67