Learn More
Polysciences, Inc. RETICULIN STAIN KIT
SDP
Catalog No. NC0679060
Encompass
Is a metal impregnation technique, where ammoniacal silver initially binds to the tissue component of interest, the reducing agent (formalin) produces a dark insoluble precipitate, then treated by toning and fixing in sodium thiosulfate.
Catalog No. NC0679060 Supplier Polysciences, Inc. Supplier No. 25094250
Is a metal impregnation technique, where ammoniacal silver initially binds to the tissue component of interest, the reducing agent (formalin) produces a dark insoluble precipitate, then treated by toning and fixing in sodium thiosulfate.