Learn More
Polysciences, Inc. RETICULIN STAIN KIT
SDP
Catalog No. NC0375423
Encompass
Identifies reticulin fibers in tissue sections liver, kidney and spleen. Reticulin is a type III collagen found in the basement membrane of many organs and provides structural integrity.
Catalog No. NC0375423 Supplier Polysciences, Inc. Supplier No. 25094500
Identifies reticulin fibers in tissue sections liver, kidney and spleen. Reticulin is a type III collagen found in the basement membrane of many organs and provides structural integrity.