Learn More
Reticulocyte Stain, 100 mL, G-Biosciences
SCI_ED
Catalog No. S001135
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
S001135 100 mL
1 options
Catalog No. S001135 Supplier G-Biosciences Supplier No. 7862249
Add to Cart
Add to Cart
Reticulocyte stain utilizes New Methylene Blue dye for the identification of reticulocytes in blood films.
  • A red blood cell containing two or more blue stained granules would be considered positive scoring.
Specifications
Quantity 100 mL
Product Type Reticulocyte Stain