Learn More
Newcomer Supply Reticulum Control Slides, 10/set
SDP
Catalog No. NC0134445
Encompass
Newcomer Supply Reticulum Control Slides are for the positive histochemical staining of reticulum fibers; regarded as specialized connective tissue fibers. Reticulum Control Slides contain sections of positive staining liver.
Catalog No. NC0134445 Supplier Newcomer Supply Supplier No. 4620A
Newcomer Supply Reticulum Control Slides are for the positive histochemical staining of reticulum fibers; regarded as specialized connective tissue fibers. Reticulum Control Slides contain sections of positive staining liver.