Learn More
Newcomer Supply Reticulum, Gordon & Sweets Stain Kit, 250 ml kit
SDP
Catalog No. NC1566831
Encompass
Newcomer Supply Reticulum, Gordon & Sweets Stain Kit procedure is a silver staining method for demonstration of reticular fibers; regarded as specialized connective tissue fibers.  This stain is useful in the differential diagnosis of certain types of tumors.
Catalog No. NC1566831 Supplier Newcomer Supply Supplier No. 9168A
Newcomer Supply Reticulum, Gordon & Sweets Stain Kit procedure is a silver staining method for demonstration of reticular fibers; regarded as specialized connective tissue fibers.  This stain is useful in the differential diagnosis of certain types of tumors.