Learn More
Diagnostic Biosystems RETICULUM STAIN KIT Reticulum Stain Kit
Catalog No. 502407521
Encompass_Preferred
This kit stains reticulum fibers in paraffin tissue sections. The reticulum fibers stain gray to black, while the nuclei and background stain pink to rose.
Catalog No. 50-240-7521 Supplier Diagnostic Biosystems Supplier No. KT031
This kit stains reticulum fibers in paraffin tissue sections. The reticulum fibers stain gray to black, while the nuclei and background stain pink to rose.