Learn More
Santa Cruz Biotechnology Retigabine N-β-D-Glucuronide Sodium Salt, 1 mg
Catalog No. NC3315400
Encompass
Alternative name :1-Deoxy-1-[[2-[(ethoxycarbonyl)amino]-5-[[(4-fluorophenyl)methyl]amino]phenyl]amino] β-D-Glucopyranuronic AcidCAS Registration Number : 191873-41-3 (free base)Molecular weight : 501.44Molecular formula : C 22 H 25 FN 3 O 8 •Na
Catalog No. NC3315400 Supplier Santa Cruz Biotechnology Supplier No. SC477046
Alternative name :1-Deoxy-1-[[2-[(ethoxycarbonyl)amino]-5-[[(4-fluorophenyl)methyl]amino]phenyl]amino] β-D-Glucopyranuronic AcidCAS Registration Number : 191873-41-3 (free base)Molecular weight : 501.44Molecular formula : C 22 H 25 FN 3 O 8 •Na