Learn More
Medchemexpress LLC HY-14649 1g Medchemexpress, Retinoic acid CAS:302-79-4 Purity:>98%
SDP
Catalog No. 502029057
Encompass_Preferred
Medchemexpress, HY-14649 1g Retinoic acid CAS:302-79-4 Retinoic acid is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis. Retinoic acid is a natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ. Retinoic acid bind to PPARβ/δ with Kd of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9057 Supplier Medchemexpress LLC Supplier No. HY146491G
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-14649 1g Retinoic acid CAS:302-79-4 Retinoic acid is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis. Retinoic acid is a natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ. Retinoic acid bind to PPARβ/δ with Kd of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.