Learn More
Sigma Aldrich Fine Chemicals Biosciences Retinoic acid, 302-79-4, MFCD00001551, 100mg
Catalog No. 501656970
Encompass
Empirical Formula (Hill Notation) C20H28O2, ≥98% (HPLC), Molecular Weight 300.44, mp 180-181 °C (lit.), powder, Synonym: ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid
Catalog No. 50-165-6970 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R2625100MG
Empirical Formula (Hill Notation) C20H28O2, ≥98% (HPLC), Molecular Weight 300.44, mp 180-181 °C (lit.), powder, Synonym: ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid