Learn More
Medchemexpress LLC Retinoic acid, 4-hydroxy- | 66592-72-1 | MFCD00872034 | >95.0% | 316.43 g/mol | C20H28O3 | 5MG
SDP
Catalog No. 5000239565
Encompass_Preferred
4-Hydroxyretinoic acid is an all-trans retinoid derivative and a metabolite of all-trans retinoic acid, used as a research reagent to study retinoid signaling, gene regulation, and cell differentiation through activation of retinoic acid receptors.
  • Research reagent for biochemical and cellular studies.
  • Metabolite of all-trans retinoic acid with retinoic acid receptor activity.
  • High purity (>95.0%) suitable for analytical applications.
  • Solid form for convenient handling and storage.
  • Molecular formula C20H28O3 and molecular weight ~316.4 g/mol.
Catalog No. 50-002-39565 Supplier Medchemexpress LLC Supplier No. HY1259045MG
4-Hydroxyretinoic acid is an all-trans retinoid derivative and a metabolite of all-trans retinoic acid, used as a research reagent to study retinoid signaling, gene regulation, and cell differentiation through activation of retinoic acid receptors.
  • Research reagent for biochemical and cellular studies.
  • Metabolite of all-trans retinoic acid with retinoic acid receptor activity.
  • High purity (>95.0%) suitable for analytical applications.
  • Solid form for convenient handling and storage.
  • Molecular formula C20H28O3 and molecular weight ~316.4 g/mol.