Learn More
Medchemexpress LLC Retinoic acid, 4-oxo-, methyl ester | 38030-58-9 | 98.8% | 328.45 g/mol | C21H28O3 | 250 MG
SDP
Catalog No. 5000285163
Encompass_Preferred
Methyl-4-oxoretinoate is a methyl ester derivative of retinoic acid used as a research reagent. It is employed in biological studies for its effects on cell signaling and skin carcinogenesis models, including inhibition of TPA-induced ornithine decarboxylase activity.
  • High purity (98.8% LCMS).
  • Molecular formula C21H28O3; molecular weight 328.45 g/mol.
  • Off-white to light yellow solid appearance.
  • Reported to inhibit TPA-induced ornithine decarboxylase activity and reduce carcinogen-induced papillomas in mouse skin.
  • Supplied as solid research material; common package sizes include 250 MG.
Catalog No. 50-002-85163 Supplier Medchemexpress LLC Supplier No. HYW364575250MG
Methyl-4-oxoretinoate is a methyl ester derivative of retinoic acid used as a research reagent. It is employed in biological studies for its effects on cell signaling and skin carcinogenesis models, including inhibition of TPA-induced ornithine decarboxylase activity.
  • High purity (98.8% LCMS).
  • Molecular formula C21H28O3; molecular weight 328.45 g/mol.
  • Off-white to light yellow solid appearance.
  • Reported to inhibit TPA-induced ornithine decarboxylase activity and reduce carcinogen-induced papillomas in mouse skin.
  • Supplied as solid research material; common package sizes include 250 MG.