Learn More
Medchemexpress LLC HY-14649 500mg , Retinoic acid CAS:302-79-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14649500MG
Encompass_Preferred
HY-14649 500mg Retinoic acid CAS: 302-79-4 Retinoic acid is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis. Retinoic acid is a natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ. Retinoic acid bind to PPARβ/δ with Kd of 17 nM. Retinoic acid acts as an inhibitor of transcription factor Nrf2 through activation of retinoic acid receptor alpha. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9521