Learn More
Sigma Aldrich Fine Chemicals Biosciences Retinoic Acid, 302-79-4, MFCD00001551, 500mg
Catalog No. NC1628240
Encompass
Molecular Formula: C20H28O2, Molecular Weight: 300.44, Assay: ≥98% (HPLC), Form: Powder, Storage Temperature: −20°C
Catalog No. NC1628240 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R2625500MG
Molecular Formula: C20H28O2, Molecular Weight: 300.44, Assay: ≥98% (HPLC), Form: Powder, Storage Temperature: −20°C