Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinoic Acid, All Trans Isomer, MP Biomedicals™

Catalog No. MP021902696 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021902696 50 mg
MP021902695 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021902696 Supplier MP Biomedicals, Inc Supplier No. 02190269.6
Add to Cart
Add to Cart

Mediates the functions of vitamin A required for growth and development. Retinoic Acid, All Trans Isomer, MP Biomedicals has been used in various differentiation protocols, including B-cells, T cells and neurons, and is applied clinically to treat cancer

Retinoic acid (RA) is a vitamin A-derived, non-peptidic, small lipophilic molecule. It affects gene transcription and modulates a wide variety of biological processes, including cell proliferation and differentiation, including apoptosis.

  • Retinoic Acid mediated gene transcription depends on the rate of transport of Retinoic Acid to target cells and the timing of exposure of Retinoic Acid to RARs (Retinoic Acid Receptors) in the target tissues.
  • All-trans-Retinoic acid (ATRA) is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR).
  • The bound RAR and RXR act as transcription factors that regulate the growth and differentiation of both normal and malignant cells.
  • Cytochromes P450 (CYPs) catalyze the 4-hydroxylation of ATRA.
  • Retinoic acid primes embryonic stem cells to become neurons.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

Melting Point 173°C to 183°C
Quantity 50 mg
Solubility Information Soluble in methanol, chloroform (40mg/mL) or acetone (30mg/mL), slightly soluble in ethanol.
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
Formula Weight 300.4
Physical Form Powder
Recommended Storage 0°C, desiccate, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.